Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium(III) carbonate hydrate, 99.9%, Thermo Scientific™

Catalog No. AC317160010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC317160010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC317160010 Supplier Thermo Scientific Chemicals Supplier No. 317160010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5895-48-7
Molecular Formula C3H6Eu2O9
Molecular Weight (g/mol) 490
InChI Key GVFRJXSZMKQNTO-UHFFFAOYSA-N
Synonym europium iii carbonate hydrate
PubChem CID 131881620
IUPAC Name carbonic acid;europium
SMILES C(=O)(O)O.C(=O)(O)O.C(=O)(O)O.[Eu].[Eu]

Specifications

Quantity 1 g
Assay Percent Range 99.9%
Formula Weight 483.95
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Europium(III) carbonate hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.