accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58965-66-5

IUPAC Name: 1,2,3,4,5-pentabromo-6-[2,3,5,6-tetrabromo-4-(2,3,4,5,6-pentabromophenoxy)phenoxy]benzene
Synonyms: 1,2,3,4,5-pentabromo-6-[2,3,5,6-tetrabromo-4-(2,3,4,5,6-pentabromophenoxy)phenoxy]benzene
Molecular Weight (g/mol): 1366.85
Molecular Formula: C18Br14O2
InChi Key: YMIUHIAWWDYGGU-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C(Br)=C(OC2=C(Br)C(Br)=C(OC3=C(Br)C(Br)=C(Br)C(Br)=C3Br)C(Br)=C2Br)C(Br)=C1Br
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results