Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetradecabromo-1,4-diphenoxybenzene [for Flame retardants], TCI America™
SDP

Catalog No. T134625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T134625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T134625G Supplier TCI America Supplier No. T134625G

Chemical Identifiers

CAS 58965-66-5
Molecular Formula C18Br14O2
Molecular Weight (g/mol) 1366.85
MDL Number MFCD00145371
InChI Key YMIUHIAWWDYGGU-UHFFFAOYSA-N
Synonym Perbromo-1,4-diphenoxybenzene
PubChem CID 100907
IUPAC Name 1,2,3,4,5-pentabromo-6-[2,3,5,6-tetrabromo-4-(2,3,4,5,6-pentabromophenoxy)phenoxy]benzene
SMILES BrC1=C(Br)C(Br)=C(OC2=C(Br)C(Br)=C(OC3=C(Br)C(Br)=C(Br)C(Br)=C3Br)C(Br)=C2Br)C(Br)=C1Br

Specifications

Color White
Quantity 25 g
Formula Weight 1366.85
Physical Form Crystalline Powder
Chemical Name or Material Tetradecabromo-1,4-diphenoxybenzene [for Flame retardants]
EINECSNumber (3)-4206
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.