CAS RN 59-14-3
5-Bromo-2'-deoxyuridine, 99%
CAS: 59-14-3 Molecular Formula: C9H11BrN2O5 Molecular Weight (g/mol): 307.10 MDL Number: MFCD00006529 InChI Key: WOVKYSAHUYNSMH-RRKCRQDMSA-N Synonym: 5-bromo-2'-deoxyuridine,broxuridine,bromodeoxyuridine,brdu,5-bromodeoxyuridine,5-brdu,budr,5-bromouracil deoxyriboside,bromouracil deoxyriboside,5-bromodesoxyuridine PubChem CID: 6035 ChEBI: CHEBI:472552 IUPAC Name: 5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione SMILES: OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O
Thermo Scientific Chemicals 5-Bromo-2'-deoxyuridine, 99+%
CAS: 59-14-3 Molecular Formula: C9H11BrN2O5 Molecular Weight (g/mol): 307.10 MDL Number: MFCD00006529 InChI Key: WOVKYSAHUYNSMH-RRKCRQDMSA-N Synonym: 5-bromo-2'-deoxyuridine,broxuridine,bromodeoxyuridine,brdu,5-bromodeoxyuridine,5-brdu,budr,5-bromouracil deoxyriboside,bromouracil deoxyriboside,5-bromodesoxyuridine PubChem CID: 6035 ChEBI: CHEBI:472552 IUPAC Name: 5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione SMILES: OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O
5-Bromo-2'-Deoxyuridine, >99%, MP Biomedicals™
CAS: 59-14-3 Molecular Formula: C9H11BrN2O5 Molecular Weight (g/mol): 307.10 MDL Number: MFCD00006529 InChI Key: WOVKYSAHUYNSMH-RRKCRQDMSA-N Synonym: 5-bromo-2'-deoxyuridine,broxuridine,bromodeoxyuridine,brdu,5-bromodeoxyuridine,5-brdu,budr,5-bromouracil deoxyriboside,bromouracil deoxyriboside,5-bromodesoxyuridine PubChem CID: 6035 ChEBI: CHEBI:472552 IUPAC Name: 5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione SMILES: OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O
MilliporeSigma™ Calbiochem™ 5-Bromo-2'-deoxyuridine
Can be used both in vitro and in vivo to label DNA during DNA synthesis where it is incorporated into DNA in place of thymidine
5-Bromo-2'-Deoxyuridine, MP Biomedicals™
CAS: 59-14-3 Molecular Formula: C9H11BrN2O5 Molecular Weight (g/mol): 307.10 MDL Number: MFCD00006529 InChI Key: WOVKYSAHUYNSMH-RRKCRQDMSA-N Synonym: 5-bromo-2'-deoxyuridine,broxuridine,bromodeoxyuridine,brdu,5-bromodeoxyuridine,5-brdu,budr,5-bromouracil deoxyriboside,bromouracil deoxyriboside,5-bromodesoxyuridine PubChem CID: 6035 ChEBI: CHEBI:472552 IUPAC Name: 5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione SMILES: OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O
5-Bromo-2'-deoxyuridine 98.0+%, TCI America™
CAS: 59-14-3 Molecular Formula: C9H11BrN2O5 Molecular Weight (g/mol): 307.10 MDL Number: MFCD00006529 InChI Key: WOVKYSAHUYNSMH-RRKCRQDMSA-N Synonym: 5-bromo-2'-deoxyuridine,broxuridine,bromodeoxyuridine,brdu,5-bromodeoxyuridine,5-brdu,budr,5-bromouracil deoxyriboside,bromouracil deoxyriboside,5-bromodesoxyuridine PubChem CID: 6035 ChEBI: CHEBI:472552 IUPAC Name: 5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione SMILES: OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O
5-Bromo-2'-deoxyuridine, Fisher BioReagents
CAS: 59-14-3 Molecular Formula: C9H11BrN2O5 Molecular Weight (g/mol): 307.10 MDL Number: MFCD00006529 InChI Key: WOVKYSAHUYNSMH-RRKCRQDMSA-N Synonym: 5-bromo-2'-deoxyuridine,broxuridine,bromodeoxyuridine,brdu,5-bromodeoxyuridine,5-brdu,budr,5-bromouracil deoxyriboside,bromouracil deoxyriboside,5-bromodesoxyuridine PubChem CID: 6035 ChEBI: CHEBI:472552 IUPAC Name: 5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione SMILES: OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O