Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2'-deoxyuridine, Fisher BioReagents

Catalog No. BP25081 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP25081 1 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP25081

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

5-Bromo-2'-Deoxyuridine is a thymidine analog used as a mutagen in genetic research.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-14-3
Molecular Formula C9H11BrN2O5
Molecular Weight (g/mol) 307.10
MDL Number MFCD00006529
InChI Key WOVKYSAHUYNSMH-RRKCRQDMSA-N
Synonym 5-bromo-2'-deoxyuridine, broxuridine, bromodeoxyuridine, brdu, 5-bromodeoxyuridine, 5-brdu, budr, 5-bromouracil deoxyriboside, bromouracil deoxyriboside, 5-bromodesoxyuridine
PubChem CID 6035
ChEBI CHEBI:472552
IUPAC Name 5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O

Specifications

Melting Point 191°C
Color White
Infrared Spectrum Conforms to reference
Assay Percent Range >99%
Packaging Amber Glass
Quantity 1 g
Merck Index 15, 1461
Formula Weight 307.1
Physical Form Powder or Solid
Chemical Name or Material 5-Bromo-2'-Deoxyuridine, Off-white Powder
Safety handling Check Alert storage code Red
Hazard Category Signal Word: WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes and skin. Possible risks of irreversible effects.
NFPA
Health:1 Flammability:0 Instability:0
Recommended Storage -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.