accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 590-14-7

IUPAC Name: (1E)-1-bromoprop-1-ene
Synonyms: (1E)-1-bromoprop-1-ene
Molecular Weight (g/mol): 120.98
Molecular Formula: C3H5Br
InChi Key: NNQDMQVWOWCVEM-NSCUHMNNSA-N
SMILES: C\C=C\Br
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)