accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3H3C

CAS RN 590-35-2

IUPAC Name: 2,2-dimethylpentane
Synonyms: 2,2-dimethylpentane
Molecular Weight (g/mol): 100.21
Molecular Formula: C7H16
InChi Key: CXOWYJMDMMMMJO-UHFFFAOYSA-N
SMILES: CCCC(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)