accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 590-63-6

ClH3CNH3CH3CCH3ONH2O

CAS RN 590-63-6

IUPAC Name: 1-(trimethylazaniumyl)propan-2-yl carbamate chloride
Synonyms: bethanechol chloride 氯贝胆碱
Molecular Weight (g/mol): 196.68
Molecular Formula: C7H17ClN2O2
InChi Key: XXRMYXBSBOVVBH-UHFFFAOYNA-N
SMILES: [Cl-].CC(C[N+](C)(C)C)OC(N)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)