Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bethanechol Chloride 98.0+%, TCI America™
SDP

Catalog No. B48285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48285G 5 g
B482825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48285G Supplier TCI America Supplier No. B48285G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 590-63-6
Molecular Formula C7H17ClN2O2
Molecular Weight (g/mol) 196.675
MDL Number MFCD00055224
InChI Key XXRMYXBSBOVVBH-UHFFFAOYSA-N
Synonym bethanechol chloride, urecholine, myocholine, besacholine, mechotane, mechothane, mecothane, myotonachol, myotonine, mictone
PubChem CID 11548
ChEBI CHEBI:3085
IUPAC Name 2-carbamoyloxypropyl(trimethyl)azanium;chloride
SMILES CC(C[N+](C)(C)C)OC(=O)N.[Cl-]

Specifications

Melting Point 216°C
Color White
Quantity 5 g
Formula Weight 196.68
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bethanechol Chloride
RTECSNumber BR5425000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.