accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59044-29-0

IUPAC Name: (9Z,12Z,15Z)-octadeca-9,12,15-trienoyl chloride
Synonyms: (9Z,12Z,15Z)-octadeca-9,12,15-trienoyl chloride
Molecular Weight (g/mol): 296.88
Molecular Formula: C18H29ClO
InChi Key: MRKXCQPDRPTZCG-PDBXOOCHSA-N
SMILES: CC\C=C/C\C=C/C\C=C/CCCCCCCC(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results