Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Linolenoyl chloride 70%, tech., Thermo Scientific™

Catalog No. AC413210010
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC413210010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC413210010 Supplier Thermo Scientific Chemicals Supplier No. 413210010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 59044-29-0
Assay Percent Range 70%
Molecular Formula C18H29ClO
Linear Formula CH3(CH2CHCH)3(CH2)7COCl
Quantity 1g
MDL Number MFCD00041727
Synonym linolenoyl chloride, 9z,12z,15z-octadeca-9,12,15-trienoyl chloride, 9,12,15-octadecatrienoyl chloride, 9z,12z,15z, 9,12,15-octadecatrienoyl chloride, z,z,z, alpha-linolenic acid chloride, 9z,12z,15z-9,12,15-octadecatrienoyl chloride, 9z,12z,15z-1-oxo-9,12,15-octadecatrienyl chloride
InChI Key MRKXCQPDRPTZCG-PDBXOOCHSA-N
SMILES CCC=CCC=CCC=CCCCCCCCC(=O)Cl
IUPAC Name (9Z,12Z,15Z)-octadeca-9,12,15-trienoyl chloride
Molecular Weight (g/mol) 296.879
PubChem CID 6436844
Formula Weight 296.88
Grade Technical
Chemical Name or Material Linolenoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
In contact with water releases flammable gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.