accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5908-27-0

IUPAC Name: 3,4-dioxo-3,4-dihydronaphthalene-1-sulfonic acid potassium
Synonyms: 3,4-dioxonaphthalene-1-sulfonic acid potassium
Molecular Weight (g/mol): 277.31
Molecular Formula: C10H6KO5S
InChi Key: UNZQHCMHYSYLOA-UHFFFAOYSA-N
SMILES: [K].OS(=O)(=O)C1=CC(=O)C(=O)C2=CC=CC=C12
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results