CAS RN 591-01-5
IUPAC Name:
bis((diaminomethylidene)urea); sulfuric acid
Synonyms:
bis(diaminomethylideneurea); sulfuric acid
Molecular Weight (g/mol):
302.27
Molecular Formula:
C4H14N8O6S
InChi Key:
IATXFPUBPMZBPH-UHFFFAOYSA-N
SMILES:
OS(O)(=O)=O.NC(N)=NC(N)=O.NC(N)=NC(N)=O
1
–
3
of
3
results
1
Promotions Available
Guanylurea sulfate, 97%
Guanylurea sulfate, 97%
CAS: 591-01-5 Molecular Formula: C4H12N8O2·H2SO4 Molecular Weight (g/mol): 302.26 MDL Number: MFCD00035565
2
Promotions Available
N-Guanylurea sulfate, 97%
N-Guanylurea sulfate, 97%
CAS: 591-01-5 Molecular Formula: C4H12N8O2·H2SO4 MDL Number: MFCD00035565 Synonym: Dicyanodiamidine sulfate
3
Guanylurea Sulfate 98.0+%, TCI America™
CAS: 591-01-5 Molecular Formula: C2H6N4O MDL Number: MFCD00035565 Synonym: Dicyandiamidine Sulfate