Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanylurea Sulfate 98.0+%, TCI America™
SDP

Catalog No. D0433500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0433500G 500 g
D043325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0433500G Supplier TCI America Supplier No. D0433500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 591-01-5
Molecular Formula C2H6N4O
MDL Number MFCD00035565
Synonym Dicyandiamidine Sulfate

Specifications

Melting Point 190°C
Color White
Quantity 500 g
Formula Weight 302.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Guanylurea Sulfate
EINECSNumber (2)-1778
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.