accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3C

CAS RN 591-21-9

IUPAC Name: 1,3-dimethylcyclohexane
Synonyms: 1,3-dimethylcyclohexane 1,3-dimethylcyclohexane (cis)
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: SGVUHPSBDNVHKL-UHFFFAOYNA-N
SMILES: CC1CCCC(C)C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results