CAS RN 591-51-5
IUPAC Name:
lithium(1+) benzenide
Synonyms:
phenyllithium
Molecular Weight (g/mol):
84.05
Molecular Formula:
C6H5Li
InChi Key:
DBYQHFPBWKKZAT-UHFFFAOYSA-N
SMILES:
[Li+].C1=CC=[C-]C=C1
1
–
1
of
1
results
1
Sigma Aldrich Phenyllithium Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.