Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenyllithium Solution

Catalog No. 593230800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
100 mL
800 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-323-0800ML 800 mL
59-323-050ML 50 mL
59-323-0100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 59-323-0800ML Supplier Sigma Aldrich Supplier No. 593230800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phenyllithium solution can be used to synthesize:1-Acetyl-1'sa-diphenylphosphinoferrocene from ferrocenophane.Ethyl 2-acetylimino-1-benzyl-6-bromo-4-phenyl-1,4-dihydroquinoline-3-carboxylate from ethyl 1-benzyl-6-bromo-2-imino-1,2-dihydroquinoline-3-carboxylate.Ethyl 2-amino-1-benzoyl-6-bromo-4-phenyl-1,4-dihydroquinoline-3-carboxylate from (E)-ethyl 2-(acetylimino)-1-benzoyl-6-bromo-1,2-dihydroquinoline-3-carboxylate.

Specifications

CAS 591-51-5
Density 0.835 g/mL at 25°C
Molecular Formula C6H5Li
Linear Formula C6H5Li
MDL Number MFCD00014254
Quantity 800 mL
Synonym PhLi
Molecular Weight (g/mol) 84.04 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.