accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3

CAS RN 591-76-4

IUPAC Name: 2-methylhexane
Synonyms: 2-methylhexane
Molecular Weight (g/mol): 100.21
Molecular Formula: C7H16
InChi Key: GXDHCNNESPLIKD-UHFFFAOYSA-N
SMILES: CCCCC(C)C
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)