accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 591-97-9

IUPAC Name: (2E)-1-chlorobut-2-ene
Synonyms: 2-butene, 1-chloro-, (E)-
Molecular Weight (g/mol): 90.55
Molecular Formula: C4H7Cl
InChi Key: YTKRILODNOEEPX-NSCUHMNNSA-N
SMILES: C\C=C\CCl
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)