Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Crotyl chloride

Catalog No. 28115100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-115-100ML 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 28-115-100ML Supplier Sigma Aldrich Supplier No. 28115100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Crotyl chloride

Specifications

CAS 591-97-9
Density 0.923 g/mL (at 20°C (literature))
Boiling Point 85°C to 86°C (lit.)
Molecular Formula C4H7Cl
Refractive Index n20/D 1.430
Linear Formula CH3CH=CHCH2Cl
MDL Number MFCD00000987
Quantity 25 mg
Synonym cis,trans-1-Chloro-2-butene
Molecular Weight (g/mol) 90.55
Percent Purity ∼70% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.