accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O(E)(E)

CAS RN 5910-87-2

IUPAC Name: (2E,4E)-nona-2,4-dienal
Synonyms: trans, trans-nona-2,4-dienal
Molecular Weight (g/mol): 138.21
Molecular Formula: C9H14O
InChi Key: ZHHYXNZJDGDGPJ-BSWSSELBSA-N
SMILES: CCCC\C=C\C=C\C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results