Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-2,4-Nonadienal 85.0+%, TCI America™
SDP

Catalog No. N05215ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05215ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N05215ML Supplier TCI America Supplier No. N05215ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 5910-87-2
Color Yellow
Boiling Point 99°C
Molecular Formula C9H14O
Quantity 5 mL
MDL Number MFCD00007006
Synonym 2,4-nonadienal, trans,trans-2,4-nonadienal, 2e,4e-nona-2,4-dienal, nonadienal, 2,4-nonadienal, e,e, 2,4-nonadienal, 2e,4e, 2,4-trans,trans-nonadienal, trans,trans-nona-2,4-dienal, unii-y411g77sh7, trans,trans-2,4-nonadien-1-al
InChI Key ZHHYXNZJDGDGPJ-BSWSSELBSA-N
SMILES CCCC\C=C\C=C\C=O
IUPAC Name (2E,4E)-nona-2,4-dienal
Molecular Weight (g/mol) 138.21
PubChem CID 5283339
Formula Weight 138.21
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material trans,trans-2,4-Nonadienal
Show More Show Less
EINECSNumber (2)-2462
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.