accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OHO

CAS RN 59115-26-3

IUPAC Name: {2-[2-(dimethylamino)ethoxy]phenyl}methanol
Synonyms: {2-[2-(dimethylamino)ethoxy]phenyl}methanol
Molecular Weight (g/mol): 195.26
Molecular Formula: C11H17NO2
InChi Key: UNABPRGGCSJBPG-UHFFFAOYSA-N
SMILES: CN(C)CCOC1=CC=CC=C1CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results