accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2NH3CNOO

CAS RN 59146-96-2

IUPAC Name: 2,3-dimethyl-6-nitroaniline
Synonyms: 2,3-dimethyl-6-nitroaniline
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: YEFYPFWBLCARLC-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(N)=C1C)[N+]([O-])=O
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)