Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Nitro-2,3-xylidine 98.0+%, TCI America™
SDP

Catalog No. N0500500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0500500G 500 g
N050025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0500500G Supplier TCI America Supplier No. N0500500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 59146-96-2 | C8H10N2O2 | 166.18 g/mol

Chemical Identifiers

CAS 59146-96-2
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00010148
InChI Key YEFYPFWBLCARLC-UHFFFAOYSA-N
PubChem CID 597166
IUPAC Name 2,3-dimethyl-6-nitroaniline
SMILES CC1=C(C(=C(C=C1)[N+](=O)[O-])N)C

Specifications

Melting Point 118°C
Quantity 500 g
Physical Form Crystal-Powder at 20°C
UN Number 2811
Synonym 6-nitro-2,3-xylidine, 2,3-dimethyl-6-nitro-aniline, 2,3-dimethyl-6-nitrophenylamine, 4-nitro-3-aminoxylol, acmc-209mau, benzenamine,2,3-dimethyl-6-nitro, benzenamine, 2,3-dimethyl-6-nitro
Formula Weight 166.18
Percent Purity ≥98.0% (GC)
Grade Not Available
Chemical Name or Material 6-Nitro-2,3-xylidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.