accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 592-55-2

IUPAC Name: 1-bromo-2-ethoxyethane
Synonyms: ethane, 1-bromo-2-ethoxy-
Molecular Weight (g/mol): 153.02
Molecular Formula: C4H9BrO
InChi Key: MMYKTRPLXXWLBC-UHFFFAOYSA-N
SMILES: CCOCCBr
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (6)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)