accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5921-80-2

IUPAC Name: 1,1-dibutoxybutane
Synonyms: 1,1-dibutoxybutane
Molecular Weight (g/mol): 202.34
Molecular Formula: C12H26O2
InChi Key: WQRBJFZKPWPIJD-UHFFFAOYSA-N
SMILES: CCCCOC(CCC)OCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results