accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOHCH3NH3CCH3

CAS RN 59229-09-3

IUPAC Name: 2,4,6-trimethylpyridine; 4-methylbenzene-1-sulfonic acid
Synonyms: collidinium p-toluenesulfonate
Molecular Weight (g/mol): 293.38
Molecular Formula: C15H19NO3S
InChi Key: VEXWNPGPVMYVDU-UHFFFAOYSA-N
SMILES: CC1=CC(C)=NC(C)=C1.CC1=CC=C(C=C1)S(O)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results