Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethylpyridinium p-Toluenesulfonate 98.0+%, TCI America™
SDP

Catalog No. T112225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T112225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T112225G Supplier TCI America Supplier No. T112225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59229-09-3
Molecular Formula C15H19NO3S
MDL Number MFCD00012810
Synonym 2,4,6-Collidinium p-Toluenesulfonate, CPTS

Specifications

Melting Point 130°C
Quantity 25 g
Formula Weight 293.38
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trimethylpyridinium p-Toluenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.