accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2N

CAS RN 5925-93-9

IUPAC Name: 2-amino-5-methylbenzonitrile
Synonyms: 2-amino-5-methylbenzonitrile
Molecular Weight (g/mol): 132.17
Molecular Formula: C8H8N2
InChi Key: OZLMBXPYRDASTP-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C(=C1)C#N
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)