Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-methylbenzonitrile 97.0+%, TCI America™
SDP

Catalog No. A28431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A28431G Supplier TCI America Supplier No. A28431G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5925-93-9
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.166
MDL Number MFCD00267506
InChI Key OZLMBXPYRDASTP-UHFFFAOYSA-N
Synonym 2-amino-5-methyl-benzonitrile, benzonitrile, 2-amino-5-methyl, 2-amino-5-methylbenzonitril, 6-amino-m-tolunitrile, pubchem17447, 2-cyano-4-methylaniline, acmc-209ww6, 2-amino-5-methyl benzonitrile, benzonitrile,2-amino-5-methyl, 2-amino-5-methylbenzonitrile
PubChem CID 242778
IUPAC Name 2-amino-5-methylbenzonitrile
SMILES CC1=CC(=C(C=C1)N)C#N

Specifications

Melting Point 60°C
Boiling Point 152°C
Quantity 1 g
UN Number 3439
Formula Weight 132.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-methylbenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.