accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5926-51-2

IUPAC Name: 3-bromo-2,5-dihydrofuran-2,5-dione
Synonyms: 3-bromofuran-2,5-dione
Molecular Weight (g/mol): 176.95
Molecular Formula: C4HBrO3
InChi Key: YPRMWCKXOZFJGF-UHFFFAOYSA-N
SMILES: BrC1=CC(=O)OC1=O
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)