accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 593-49-7

IUPAC Name: heptacosane
Synonyms: heptacosane
Molecular Weight (g/mol): 380.75
Molecular Formula: C27H56
InChi Key: BJQWYEJQWHSSCJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCC
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (5)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)