accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 593-85-1

IUPAC Name: carbonic acid; bis(guanidine)
Synonyms: carbonic acid, compd. with guanidine (1:2)
Molecular Weight (g/mol): 180.17
Molecular Formula: C3H12N6O3
InChi Key: STIAPHVBRDNOAJ-UHFFFAOYSA-N
SMILES: NC(N)=N.NC(N)=N.OC(O)=O
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)