Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine carbonate, 98%

Catalog No. AAA1285330
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1285330 250 g
AAA128530B 1000 g
AAA128530I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1285330 Supplier Thermo Scientific Chemicals Supplier No. A1285330
  • $40.60
    $38.65 / Each of 1

    Save $1.95

    5% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 593-85-1 | C3H12N6O3

It is employed as a strong organic alkali, organic intermediate, in soap and cosmetic products, and in the textile industry. It is also used to make fine chemicals. It is used in various industries to solve different purpose such as detergents as a synergistic agent, pharmaceuticals as an antibiotic and microencapsulation process for carbonless paper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a strong organic alkali, organic intermediate, in soap and cosmetic products, and in the textile industry. It is also used to make fine chemicals. It is used in various industries to solve different purpose such as detergents as a synergistic agent, pharmaceuticals as an antibiotic and microencapsulation process for carbonless paper.

Solubility
Soluble in water 450 g/L (20°C); slightly soluble in alcohol and acetone.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-85-1
Molecular Formula C3H12N6O3
MDL Number MFCD00013029

Specifications

Melting Point ∼198°C (decomposition)
Quantity 250 g
Beilstein 3628359
Solubility Information Soluble in water 450g/L (20°C); slightly soluble in alcohol and acetone.
Formula Weight 180.17
Percent Purity 98%
Chemical Name or Material Guanidine carbonate
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 209-813-7
RTECSNumber FG1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.