accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHONH2NH2HN

CAS RN 593-87-3

IUPAC Name: acetic acid; guanidine
Synonyms: acetic acid; guanidine
Molecular Weight (g/mol): 119.12
Molecular Formula: C3H9N3O2
InChi Key: DXTIKTAIYCJTII-UHFFFAOYSA-N
SMILES: CC(O)=O.NC(N)=N
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results