Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine acetate, 99%, Thermo Scientific™

Catalog No. AC268990250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC268990250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC268990250 Supplier Thermo Scientific Chemicals Supplier No. 268990250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 593-87-3
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C3H9N3O2
Linear Formula NH2C(NH)nH2·CH3COOH
Quantity 25g
Beilstein 03, 152
Synonym guanidine acetate, guanidine, monoacetate, guanidine acetate salt, acetic acid; guanidine, guanidine, acetate 1:1, guanidine monoacetate, guanidinium monoacetate, acetic acid-guanidine 1/1, amino imino methanaminium acetate
InChI Key DXTIKTAIYCJTII-UHFFFAOYSA-N
SMILES CC(=O)O.C(=N)(N)N
IUPAC Name acetic acid;guanidine
Molecular Weight (g/mol) 119.124
PubChem CID 2724684
Formula Weight 119.12
Percent Purity ≥98.5%
Chemical Name or Material Guanidine acetate, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-814-2
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.