accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH3N

CAS RN 5930-98-3

IUPAC Name: 6-methoxy-2,4-dimethyl-2H,3H-furo[3,2-c]quinoline
Synonyms: 6-methoxy-2,4-dimethyl-2H,3H-furo[3,2-c]quinoline
Molecular Weight (g/mol): 229.28
Molecular Formula: C14H15NO2
InChi Key: MZPYLXKRBJRNSK-UHFFFAOYNA-N
SMILES: COC1=C2N=C(C)C3=C(OC(C)C3)C2=CC=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)