Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trimethylsilyl)-3-butyn-2-one 97.0+%, TCI America™
SDP

Catalog No. T29735ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29735ML 5 mL
T297325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29735ML Supplier TCI America Supplier No. T29735ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5930-98-3
Molecular Formula C14H15NO2
Molecular Weight (g/mol) 229.28
MDL Number MFCD00190212
InChI Key MZPYLXKRBJRNSK-UHFFFAOYNA-N
Synonym 4-trimethylsilyl-3-butyn-2-one, 4-trimethylsilyl but-3-yn-2-one, 3-butyn-2-one, 4-trimethylsilyl, pubchem16060, ksc490e2j
PubChem CID 2734664
IUPAC Name 6-methoxy-2,4-dimethyl-2H,3H-furo[3,2-c]quinoline
SMILES COC1=C2N=C(C)C3=C(OC(C)C3)C2=CC=C1

Specifications

Color Yellow
Boiling Point 158°C
Quantity 5 mL
UN Number 1993
Formula Weight 140.26
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Trimethylsilyl)-3-butyn-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.