accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNSO

CAS RN 59303-07-0

IUPAC Name: 2-{[(furan-2-yl)methyl]sulfanyl}-3-methylpyrazine
Synonyms: 2-[(furan-2-ylmethyl)sulfanyl]-3-methylpyrazine
Molecular Weight (g/mol): 206.26
Molecular Formula: C10H10N2OS
InChi Key: PFRSWMCUERVSAT-UHFFFAOYSA-N
SMILES: CC1=NC=CN=C1SCC1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results