Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Furfurylthio-3-methylpyrazine 95.0+%, TCI America™
SDP

Catalog No. F02225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F02225G Supplier TCI America Supplier No. F02225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59303-07-0
Molecular Formula C10H10N2OS
Molecular Weight (g/mol) 206.26
MDL Number MFCD00055031
InChI Key PFRSWMCUERVSAT-UHFFFAOYSA-N
PubChem CID 101003
IUPAC Name 2-{[(furan-2-yl)methyl]sulfanyl}-3-methylpyrazine
SMILES CC1=NC=CN=C1SCC1=CC=CO1

Specifications

Quantity 5 g
Formula Weight 206.26
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Furfurylthio-3-methylpyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.