accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5933-87-9

IUPAC Name: pentyl decanoate
Synonyms: pentyl decanoate
Molecular Weight (g/mol): 242.4
Molecular Formula: C15H30O2
InChi Key: BGYFCELTJPRWBA-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(=O)OCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results