Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyl Decanoate 93.0+%, TCI America™
SDP

Catalog No. D001825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D001825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D001825ML Supplier TCI America Supplier No. D001825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5933-87-9
Molecular Formula C15H30O2
Molecular Weight (g/mol) 242.403
MDL Number MFCD00059454
InChI Key BGYFCELTJPRWBA-UHFFFAOYSA-N
Synonym Decanoic Acid Amyl Ester, Pentyl Decanoate, Decanoic Acid Pentyl Ester
PubChem CID 550768
IUPAC Name pentyl decanoate
SMILES CCCCCCCCCC(=O)OCCCCC

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 242.40
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Amyl Decanoate
EINECSNumber (2)-0782&(2)-0798
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.