CAS RN 59333-67-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Fluoxetine, Hydrochloride
A cell-permeable phenylpropylamine derived anti-depressant that acts as a selective serotonin re-uptake inhibitor (SSRI).