Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Fluoxetine, Hydrochloride

Catalog No. 34329025MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-329-025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-329-025MG Supplier MilliporeSigma™ Supplier No. 34329025MG
Add to Cart
Add to Cart

A cell-permeable phenylpropylamine derived anti-depressant that acts as a selective serotonin re-uptake inhibitor (SSRI).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Description 98% by HPLC
Target Proteins Selective serotonin re-uptake inhibitor
CAS 59333-67-4
Molecular Formula C17H18F3NO·HCL
Formula Weight 345.8g/mol
Solubility MeOH or H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.