accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59346-56-4

IUPAC Name: 3-hydroxy-2,2,4-trimethylpentanenitrile
Synonyms: 3-hydroxy-2,2,4-trimethylpentanenitrile
Molecular Weight (g/mol): 141.21
Molecular Formula: C8H15NO
InChi Key: YQUDZXMXULHYPN-UHFFFAOYNA-N
SMILES: CC(C)C(O)C(C)(C)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results