Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-2,2,4-trimethylvaleronitrile 95.0+%, TCI America™
SDP

Catalog No. T11205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T11205G Supplier TCI America Supplier No. T11205G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59346-56-4
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.214
MDL Number MFCD00059590
InChI Key YQUDZXMXULHYPN-UHFFFAOYSA-N
Synonym 2,2,4-Trimethyl-3-hydroxyvaleronitrile
PubChem CID 550070
IUPAC Name 3-hydroxy-2,2,4-trimethylpentanenitrile
SMILES CC(C)C(C(C)(C)C#N)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 141.21
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Hydroxy-2,2,4-trimethylvaleronitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.