accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NH2HNNH2NH2HNOSOOO

CAS RN 594-14-9

IUPAC Name: bis(guanidine) sulfate
Synonyms: guanidine sulfate (2:1)
Molecular Weight (g/mol): 214.2
Molecular Formula: C2H10N6O4S
InChi Key: UYAKTBUPMOOXNW-UHFFFAOYSA-L
SMILES: NC(N)=N.NC(N)=N.[O-]S([O-])(=O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)