Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine Sulfate 99.0+%, TCI America™
SDP

Catalog No. G016625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G016625G 25 g
G0166500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G016625G Supplier TCI America Supplier No. G016625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 594-14-9
Molecular Formula CH5N3
MDL Number MFCD00013115

Specifications

Color White
Quantity 25 g
Formula Weight 216.22
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Guanidine Sulfate
EINECSNumber (2)-1773
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.