accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3CH3HO

CAS RN 594-60-5

IUPAC Name: 2,3-dimethylbutan-2-ol
Synonyms: 2,3-dimethyl-2-butanol
Molecular Weight (g/mol): 102.18
Molecular Formula: C6H14O
InChi Key: IKECULIHBUCAKR-UHFFFAOYSA-N
SMILES: CC(C)C(C)(C)O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)